missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Fused Crystal, Technical, Spectrum™ Chemical

Catalog No. 18612223
Change view
Click to view available options
Quantity:
500 g
1 kg
180 kg
Packaging:
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612223 500 g Fiber Drum
18612222 1 kg Fiber Drum
18612221 180 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612223 Supplier Spectrum Chemical Mfg Cor Supplier No. P1065500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-95-2

Specifications

CAS Max % 1
Boiling Point 182°C
Packaging Fiber Drum
Quantity 500 g
Grade Technical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.