missing translation for 'onlineSavingsMsg'
Learn More

Phenol, detached crystals, 99+%

Catalog No. AAA1576022
Change view
Click to view available options
Quantity:
100 g
500 g
5 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1576022 100 g
AAA1576036 500 g
AAA1576061 5 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1576022 Supplier Thermo Scientific Chemicals Supplier No. A1576022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-95-2 | C6H6O | 94.11 g/mol

Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A, which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers, which is used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colorings and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A, which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers, which is used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colorings and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.

Solubility
Soluble in water, alcohol, chloroform, ether, benzene, glycerol, acetone, carbon disulfide and aqueous alkali hydroxides.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong acids.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 40°C to 43°C
Density 1.071
Boiling Point 181°C
Flash Point 79°C (175°F)
Odor Sweet
Quantity 100 g
UN Number UN1671
Beilstein 969616
Sensitivity Air and light sensitive
Merck Index 14,7241
Solubility Information Soluble in water,alcohol,chloroform,ether,benzene,glycerol,acetone,carbon disulfide and aqueous alkali hydroxides.
Formula Weight 94.11
Percent Purity ≥99%
Chemical Name or Material Phenol, detached crystals
Show More Show Less
Hazard Category H301+H311+H331-H314-H335-H341-H373
Hazard Statement GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID
EINECSNumber 203-632-7
RTECSNumber SJ3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only