missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol, 5% (w/v) Solution, 4.9-5.1%, Spectrum™ Chemical
$134.76 - $596.56
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18-612-218
|
Spectrum Chemical Mfg Cor
P259500ML |
500 mL | Amber Glass Bottle |
Each for $134.76
|
|
||||
|
18-612-256
|
Spectrum Chemical Mfg Cor
P2591LTPL |
1 L | Amber Glass Bottle |
Each for $190.71
|
|
||||
|
18-612-257
|
Spectrum Chemical Mfg Cor
P2594LTPL |
4 L | Amber Glass Bottle |
Each for $596.56
|
|
||||
Chemical Identifiers
| 108-95-2 |
Specifications
| 108-95-2 | |
| 4.9 to 5.1% | |
| Amber Glass Bottle | |
| 500 mL |
| 5% | |
| Chemical Solution | |
| Colorless | |
| Phenol |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.