missing translation for 'onlineSavingsMsg'
Learn More

Phenol, 5% (w/v) Solution, 4.9-5.1%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P2591LTPL

 View more versions of this product

Catalog No. 18-612-256


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenol
5%
Chemical Solution
Colorless
108-95-2
4.9 to 5.1%
Amber Glass Bottle
1 L

Chemical Identifiers

108-95-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.