missing translation for 'onlineSavingsMsg'
Learn More

Phenformin hydrochloride, MilliporeSigma™ Supelco™

Phenformin hydrochloride is a biguanide derivative that shows anti-diabetic activity.

$56.57 - $220.08

Specifications

Packaging Glass Bottle
CAS 834-28-6
Physical Form Neat
Linear Formula C10H15N5 · HCl
MDL Number MFCD00035039
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015644
View Documents
Merck Emd Millipore
P70451G
1 g
Each for $56.57
Only null left
 
111015645
View Documents
Merck Emd Millipore
P704510G
10 g
Each for $220.08
Only null left
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

Glass Bottle
Neat
MFCD00035039
Analytical Standard
Analytical Standard
834-28-6
C10H15N5 · HCl
N-(2-Phenylethyl)imidodicarbonimidic diamide monohydrochloride; Phenethylbiguanide
241.72
Videos
SDS
Documents

Documents