missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenformin hydrochloride, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Packaging | Glass Bottle |
| CAS | 834-28-6 |
| Physical Form | Neat |
| Linear Formula | C10H15N5 · HCl |
| MDL Number | MFCD00035039 |
| Synonym | N-(2-Phenylethyl)imidodicarbonimidic diamide monohydrochloride; Phenethylbiguanide |
| Type | Analytical Standard |
| Quantity | 10 g |
| Formula Weight | 241.72 |
| Grade | Analytical Standard |