missing translation for 'onlineSavingsMsg'
Learn More

Phenetole 98.0+%, TCI America™

Supplier:  TCI America E0043500ML

 View more versions of this product

Catalog No. E0043500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenetole
-30°C
173°C
MFCD00009090
3271
DLRJIFUOBPOJNS-UHFFFAOYSA-N
ethoxybenzene
7674
122.17
Liquid
103-73-1
Colorless
C8H10O
500 mL
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
CCOC1=CC=CC=C1
122.167
CHEBI:67129
≥98.0% (GC)

Chemical Identifiers

103-73-1
122.167
DLRJIFUOBPOJNS-UHFFFAOYSA-N
7674
ethoxybenzene
C8H10O
MFCD00009090
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
CHEBI:67129
CCOC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0557

RTECSNumber : SI7700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions