missing translation for 'onlineSavingsMsg'
Learn More

Phenetole 98.0+%, TCI America™

Catalog No. p-7128803
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
E004325ML 25 mL
E0043500ML 500 mL
2 options

Catalog No. E004325ML

Supplier: TCI America E004325ML

Only null left

Chemical Identifiers

CAS 103-73-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00009090
InChI Key DLRJIFUOBPOJNS-UHFFFAOYSA-N
Synonym phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
PubChem CID 7674
ChEBI CHEBI:67129
IUPAC Name ethoxybenzene
SMILES CCOC1=CC=CC=C1

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 173°C
Quantity 25 mL
UN Number 3271
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenetole
EINECSNumber (3)-0557
RTECSNumber SI7700000
TSCA Yes