Learn More
Phenetole, 98+%
CAS: 103-73-1 | C8H10O | 122.167 g/mol
Supplier: Thermo Scientific Chemicals A155010F
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Phenetole | |
| -30°C | |
| 169°C to 170°C | |
| Aromatic | |
| 1.507 | |
| 2500 mL | |
| 636270 | |
| phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f | |
| CCOC1=CC=CC=C1 | |
| 122.167 | |
| CHEBI:67129 | |
| ≥98% |
| 103-73-1 | |
| 0.967 | |
| 57°C (134°F) | |
| C8H10O | |
| MFCD00009090 | |
| UN3271 | |
| 14,7229 | |
| DLRJIFUOBPOJNS-UHFFFAOYSA-N | |
| ethoxybenzene | |
| 7674 | |
| 122.17 |
Chemical Identifiers
| 103-73-1 | |
| 122.167 | |
| DLRJIFUOBPOJNS-UHFFFAOYSA-N | |
| 7674 | |
| ethoxybenzene |
| C8H10O | |
| MFCD00009090 | |
| phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f | |
| CHEBI:67129 | |
| CCOC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 203-139-7
RTECSNumber : SI7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only