missing translation for 'onlineSavingsMsg'
Learn More

Phenetole, 98+%

Catalog No. p-7022632
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15501AE 100 mL
AAA15501AP 500 mL
AAA155010F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA15501AE Supplier Thermo Scientific Chemicals Supplier No. A15501AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-73-1 | C8H10O | 122.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-73-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00009090
InChI Key DLRJIFUOBPOJNS-UHFFFAOYSA-N
Synonym phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
PubChem CID 7674
ChEBI CHEBI:67129
IUPAC Name ethoxybenzene
SMILES CCOC1=CC=CC=C1

Specifications

Melting Point -30°C
Density 0.967
Boiling Point 169°C to 170°C
Flash Point 57°C (134°F)
Odor Aromatic
Refractive Index 1.507
Quantity 100 mL
UN Number UN3271
Beilstein 636270
Merck Index 14,7229
Formula Weight 122.17
Percent Purity ≥98%
Chemical Name or Material Phenetole
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 203-139-7
RTECSNumber SI7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only