missing translation for 'onlineSavingsMsg'
Learn More

Phenanthro[1,2-b:8,7-b']dithiophene 98.0+%, TCI America™

Catalog No. P2383100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2383100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2383100MG Supplier TCI America Supplier No. P2383100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1491133-64-2
Molecular Formula C18H10S2
Molecular Weight (g/mol) 290.398
InChI Key CFRRRFIZJNHSLM-UHFFFAOYSA-N
PubChem CID 102198612
SMILES C1=CC2=C(C=CC3=C2C=CC4=C3SC=C4)C5=C1C=CS5

Specifications

Melting Point 240°C
Color Yellow
Quantity 100 mg
Formula Weight 290.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenanthro[1,2-b:8,7-b′]dithiophene