accessibility menu, dialog, popup

UserName

SS

CAS RN 1491133-64-2

IUPAC Name: 8,15-dithiapentacyclo[11.7.0.0²,¹⁰.0⁵,⁹.0¹⁴,¹⁸]icosa-1(20),2,4,6,9,11,13,16,18-nonaene
Synonyms: 8,15-dithiapentacyclo[11.7.0.0²,¹⁰.0⁵,⁹.0¹⁴,¹⁸]icosa-1(20),2,4,6,9,11,13,16,18-nonaene
Molecular Weight (g/mol): 290.4
Molecular Formula: C18H10S2
InChi Key: CFRRRFIZJNHSLM-UHFFFAOYSA-N
SMILES: S1C=CC2=CC=C3C(C=CC4=C5SC=CC5=CC=C34)=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results