missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phénacétine 99,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 136°C |
| Color | Blanc |
| Boiling Point | 132°C |
| Quantity | 500 g |
| Formula Weight | 179.22 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Phenacetin |
Sécurité et manipulation
| EINECSNumber | (3)-0697 |
| RTECSNumber | AM4375000 |
| TSCA | Yes |