accessibility menu, dialog, popup

UserName

CH3ONHCH3O

CAS RN 62-44-2

IUPAC Name: N-(4-ethoxyphenyl)acetamide
Synonyms: phenacetin acetophenetidine acetphenetidin codempiral achrocidin fenacetina acetophenetin contradol contradouleur acetophenetidin
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: CPJSUEIXXCENMM-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(NC(C)=O)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)