missing translation for 'onlineSavingsMsg'
Learn More

Phenacetin, Powder, 98.0-101.0%, Spectrum™ Chemical

Catalog No. p-7166007
Change view
Click to view available options
Quantity:
25 kg
50 kg
Packaging:
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612209 25 kg Fiber Drum
18612210 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612209 Supplier Spectrum Chemical Mfg Cor Supplier No. P105425KGBL
Only null left

Chemical Identifiers

CAS 62-44-2

Specifications

Packaging Fiber Drum
CAS Max % 1
Melting Point 134°C to 137°C
Quantity 25 kg
Percent Purity 98 to 101%
Grade Reagent
Chemical Name or Material Phenacetin

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.