missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenacetin, Powder, 98.0-101.0%, Spectrum™ Chemical
Specifications
Specifications
| Packaging | Fiber Drum |
| CAS Max % | 1 |
| Melting Point | 134°C to 137°C |
| Quantity | 50 kg |
| Percent Purity | 98 to 101% |
| Grade | Reagent |
| Chemical Name or Material | Phenacetin |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.