Learn More
VeriSpec™ pH Buffer, Certified Reference Material
ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.
Supplier:  Ricca VeriSpec™ RV1510001A
 Afficher plus de versions de ce produit
Numéro de catalogue. RV1510001A
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • No formaldehyde or mercury compounds as preservatives
  • No chromium compounds as colorants
  • Manufactured and tested in ISO 17025/17034 accredited facilities
  • pH 4, 7, and 10 are color coded for easy identification
  • Certified traceable to NIST
Spécifications
Spécifications
7732-18-5,7778-77-0,1310-73-2,7558-79-4
Yes
pH Meter and Electrode
Liquid
1 L
1 L Natural Poly
Certified
Colorless
Calibration
6.00 ±0.01
VeriSpec
pH Buffer
ISO 17025/17034
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
ISO 17025/ISO 17034