missing translation for 'onlineSavingsMsg'
Learn More

Verispec™ PFAS Standards, Certified Reference Material

Manufactured and tested in an ISO 17025 and 17034 accredited facility for applications in environmental testing, food safety, water quality, and more.

$822.87 - $1767.60

Specifications

Quantity 100 mg
Percent Purity Lot Specific
Physical Form Solid or Liquid
View More Specs

Products 4
Catalog Number Mfr. No. Chemical Name or Material Molecular Weight (g/mol) Price Quantity  
Catalog Number Mfr. No. Chemical Name or Material Molecular Weight (g/mol) Price Quantity  
01-341-376
View Documents
Verispec™
RV020007100CM
Pentadecafluorooctanoic acid ammonium salt 431.09 g/mol
Each for $1,767.60
Only null left
 
01-341-380
View Documents
Verispec™
RV020002100CM
Perfluoroundecanoic acid 564.09 g/mol
Each for $1,365.47
Only null left
 
01-341-382
View Documents
Verispec™
RV020004100CM
Perfluorooctanic acid sodium salt 436.05 g/mol
Each for $822.87
Only null left
 
01-341-383
View Documents
Verispec™
RV020000100CM
Tricosafluorododecanoic acid 614.09 g/mol
Each for $1,646.03
Only null left
 
Description

Description

  • Available in convenient 100 mg quantities for customizable calibration standards
  • Supports environmental testing, drinking and wastewater analysis, food safety, and industrial quality control.
  • Ensure compliance with key methods:

EPA: 533, 537.1, 1633, 8327

ASTM: D7968, D7979, D8421, D8535

  • High-quality sample preparation solutions, solvents, and mobile phase reagents
  • Minimize contamination risks to enhance accuracy in sensitive applications
  • Contact Us for customs or ready to use standards
Specifications

Specifications

100 mg
Lot Specific
Solid or Liquid
Videos
SDS
Documents

Documents