missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verispec™ PFAS Standards, Certified Reference Material
Description
- Available in convenient 100 mg quantities for customizable calibration standards
- Supports environmental testing, drinking and wastewater analysis, food safety, and industrial quality control.
- Ensure compliance with key methods:
EPA: 533, 537.1, 1633, 8327
ASTM: D7968, D7979, D8421, D8535
- High-quality sample preparation solutions, solvents, and mobile phase reagents
- Minimize contamination risks to enhance accuracy in sensitive applications
- Contact Us for customs or ready to use standards
Specifications
Specifications
| Chemical Name or Material | Tricosafluorododecanoic acid |
| CAS | 307-55-1 |
| Quantity | 100 mg |
| Molecular Weight (g/mol) | 614.09 g/mol |
| Percent Purity | Lot Specific |
| Physical Form | Solid or Liquid |