missing translation for 'onlineSavingsMsg'
Learn More

Verispec™ PFAS Standards, Certified Reference Material

Catalog No. 01341376
Change view
Click to view available options
Chemical Name or Material:
Pentadecafluorooctanoic acid ammonium salt
Perfluorooctanic acid sodium salt
Perfluoroundecanoic acid
Tricosafluorododecanoic acid
Molecular Weight (g/mol):
431.09 g/mol
436.05 g/mol
564.09 g/mol
614.09 g/mol
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material Molecular Weight (g/mol)
01341376 Pentadecafluorooctanoic acid ammonium salt 431.09 g/mol
01341380 Perfluoroundecanoic acid 564.09 g/mol
01341382 Perfluorooctanic acid sodium salt 436.05 g/mol
01341383 Tricosafluorododecanoic acid 614.09 g/mol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01341376 Supplier Verispec™ Supplier No. RV020007100CM
Only null left

Manufactured and tested in an ISO 17025 and 17034 accredited facility for applications in environmental testing, food safety, water quality, and more.

  • Available in convenient 100 mg quantities for customizable calibration standards
  • Supports environmental testing, drinking and wastewater analysis, food safety, and industrial quality control.
  • Ensure compliance with key methods:

EPA: 533, 537.1, 1633, 8327

ASTM: D7968, D7979, D8421, D8535

  • High-quality sample preparation solutions, solvents, and mobile phase reagents
  • Minimize contamination risks to enhance accuracy in sensitive applications
  • Contact Us for customs or ready to use standards

Specifications

Chemical Name or Material Pentadecafluorooctanoic acid ammonium salt
CAS 3825-26-1
Quantity 100 mg
Molecular Weight (g/mol) 431.09 g/mol
Percent Purity Lot Specific
Physical Form Solid or Liquid