missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PF 06409577

Catalog No. 611450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
611450 50 mg
611410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 611450 Supplier R&D Systems™ Supplier No. 6114/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Potent and selective allosteric activator of AMPK

PF 06409577 is a potent and selective allosteric activator of AMPK (EC50 values are 7 and >40000 nM for AMPKα1β1γ1 and AMPKα1β2γ1, respectively). Exhibits minimal off-target effects on a panel of receptors, ion channels, PDEs and kinases. Improves kidney function and reduces proteinuria in a preclinical model of diabetic nephropathy. Orally bioavailable.

Specifications

Chemical Name or Material PF 06409577
CAS 1467057-23-3
Synonym 6-Chloro-5-[4-(1-hydroxycyclobutyl)phenyl]-1H-indole-3-carboxylic acid
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Product Line Tocris
Molecular Formula C19H16ClNO3
Formula Weight 341.79
Physical Form Solid
Color Gray
Product Type Bioactive Small Molecule
Purity 99%
Show More Show Less

For Research Use Only.