missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 04620110

Catalog No. 58635
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58635 5 mg
586325 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 58635 Supplier Tocris Bioscience™ Supplier No. 5863/5

Potent and selective DGAT-1 inhibitor; orally bioavailable

PF 04620110 is a potent and selective DGAT-1 inhibitor (IC50 = 19 nM). Exhibits >1000-fold selectivity for DGAT-1 over DGAT-2, and no significant activity against a range of other enzymes, ion channels and receptors. Reduces plasma triglyceride levels in a rat lipid challenge model. Orally bioavailable.

Specifications

Chemical Name or Material trans-4-[4-(4-Amino-7,8-dihydro-5-oxopyrimido[5,4-f][1,4]oxazepin-6(5H)-yl)phenyl]cyclohexaneacetic acid
CAS 1109276-89-2
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C21H24N4O4
Formula Weight Observed MW: 396.44