missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 04457845
Description
PF 04457845 is a potent and selective irreversible FAAH inhibitor (IC50 = 7.2 nM, which is selective for FAAH over a panel of other serine hydrolases. PF 04457845 exhibits efficacy in a rat inflammatory pain model, and is orally bioavailable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | N-3-Pyridazinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methylene]-1-piperidinecarboxamide |
| CAS | 1020315-31-4 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C23H20F3N5O2 |
| Formula Weight | Observed MW: 455.43 |