missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Refinery Reformate Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022554
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022554 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022554 Supplier Merck Emd Millipore Supplier No. 47489

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Petroleum Refinery Reformate is analyzed by GC-FID and GC-MS procedures. It is both qualitative and quantitative reference material for evaluating refinery process performance, identifying contamination, method development, PIANO analyses, and training.

Specifications

Packaging Ampule
Flash Point <-40°C (Closed Cup)
Physical Form Neat
UN Number UN1268 - Class 3 - PG 2 - VP > 110 kPa - Petroleum distillates, n.o.s., HI: no
Quantity 1 mL
Grade Analytical Standard