missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Honeywell™ Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, low boiling point hydrogen treated naphtha, bp≥ 90% 40-60Â °C (≥ 90%)
Supplier: Honeywell Chemicals 32299195L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| Petroleum Ether | |
| 101316-46-5 | |
| 0.640 to 0.655g/mL at 20°C | |
| 195 L | |
| Metal Drum | |
| Free acid (as CH3COOH): ≤0.002% | |
| UN1268 |
| Reag. ISO | |
| Liquid | |
| 40°C to 60°C | |
| −30°C (−22°F) | |
| 0 to 0.001% | |
| MFCD00081849 | |
| ACS/puriss. p.a. |
Chemical Identifiers
| 101316-46-5 | |
| MFCD00081849 |
Safety and Handling
P210-P261-P273-P301 + P310-P331
H225-H302-H304-H315-H336-H411
DOTInformation : Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S