CAS RN 101316-46-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
5
of
5
results
1
Petroleum Ether, puriss. p.a., Reag. Ph. Eur., Solstice
CAS: 101316-46-5 MDL Number: MFCD00081849
2
Petroleum Ether, puriss. p.a., high boiling, bp 60-80°C, Solstice
CAS: 101316-46-5 MDL Number: MFCD00081849
3
Petroleum ether, For pesticide residue analysis, low boiling point hydrogen treated naphtha, bp 40-60 °C, Solstice
CAS: 101316-46-5 MDL Number: MFCD00081849
4
Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Solstice
CAS: 101316-46-5 MDL Number: MFCD00081849
5
Petroleum Ether, puriss., Solstice
CAS: 101316-46-5 MDL Number: MFCD00081849