missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Honeywell™ Riedel-de Haën™

Catalog No. p-7120659
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
195 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046119 1 L Glass Bottle
60046120 2.5 L Glass Bottle
60046124 5 L Aluminum Bottle
60046125 6 x 1 L Glass Bottle
60046122 4 x 2.5 L Glass Bottle
60046123 4 x 5 L Aluminum Bottle
60046118 195 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60046119 Supplier Honeywell Chemicals Supplier No. 322991L
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, low boiling point hydrogen treated naphtha, bp≥ 90% 40-60 °C (≥ 90%)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Appearance Liquid
Density 0.640 to 0.655g/mL at 20°C
Boiling Point 40°C to 60°C
Quantity 1 L
Flash Point −30°C (−22°F)
Packaging Glass Bottle
Residue after Evaporation 0 to 0.001%
Titratable Acid Free acid (as CH3COOH): ≤0.002%
UN Number UN1268
Grade ACS/puriss. p.a.
Chemical Name or Material Petroleum Ether, Reag. ISO
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331
Precautionary Statement H225-H302-H304-H315-H336-H411
DOTInformation Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S