missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether/Dichloromethane, 80/20 v/v, Reagents

Catalog No. 01334297
Change view
Click to view available options
Quantity:
2.5 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334297 2.5 L
01334296 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334297 Supplier Labchem Inc Supplier No. CP1208002.5C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Petroleum Ether/Dichloromethane, 80/20 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8032-32-4 , 75-09-2

Specifications

Color Colorless
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7813
Grade Reagent
Chemical Name or Material Petroleum Ether/Dichloromethane, 80/20 v/v