missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, 35° to 60°C, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$5248.89 - $5248.89
Chemical Identifiers
| CAS | 8032-32-4 |
|---|---|
| Synonym | p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Chemical Identifiers
| 8032-32-4 | |
| p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Specifications
| 8032-32-4 | |
| −18.00°C | |
| 1L = 0.68kg | |
| BAKER ANALYZED™ ACS Reagent |
| 1 L | |
| Aluminum Bottle | |
| p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide | |
| Petroleum Ether, 35° to 60°C |