missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petrolatum, Yellow, USP, Spectrum™ Chemical
PE130, 8009-03-8
Supplier: Spectrum Chemical Mfg Cor PE13025KG
Description
Spectrum™ Chemical Petrolatum, Yellow, USP is also known as petroleum jelly and is a mixture of hydrocarbons. It is typically used as a topical ointment because of its healing properties and cosmetic purposes. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 8009-03-8 | |
| 0.001 | |
| Poly Bottle | |
| 2.5 kg | |
| WJZSZXCWMATYFX-UHFFFAOYSA-N | |
| 1,1,2-trimethyl-1H-benzo[e]indole | |
| USP |
| 1 | |
| 38-60°C | |
| C15H15N | |
| 0.815-0.880 | |
| CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C | |
| 209.29 |
Chemical Identifiers
| 8009-03-8 | |
| 209.29 | |
| 1,1,2-trimethyl-1H-benzo[e]indole |
| C15H15N | |
| WJZSZXCWMATYFX-UHFFFAOYSA-N | |
| CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C |