missing translation for 'onlineSavingsMsg'
Learn More

Petrolatum, Yellow, USP, Spectrum™ Chemical

Catalog No. p-7157967
Change view
Click to view available options
Quantity:
500 g
2.5 kg
15.9 kg
175 kg
Packaging:
Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605051 500 g Poly Bottle
18605052 2.5 kg Poly Bottle
18605053 15.9 kg Poly Pail
18605054 175 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605051 Supplier Spectrum Chemical Mfg Cor Supplier No. PE130500GM
Only null left

PE130, 8009-03-8

Spectrum™ Chemical Petrolatum, Yellow, USP is also known as petroleum jelly and is a mixture of hydrocarbons. It is typically used as a topical ointment because of its healing properties and cosmetic purposes. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8009-03-8
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.29
InChI Key WJZSZXCWMATYFX-UHFFFAOYSA-N
IUPAC Name 1,1,2-trimethyl-1H-benzo[e]indole
SMILES CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C

Specifications

CAS Max % 1
Ignition Residue 0.001
Melting Range 38-60°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 0.815-0.880
Grade USP