missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Peroxidase, horseradish

Used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody.

$228.07 - $1,493.74

Specifications

Form Lyophilized
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60026MC
View Documents Promotion Details
Thermo Scientific Chemicals
J60026MC
100 mg
Each for $228.07
Only null left
 
AAJ6002603
View Documents Promotion Details
Thermo Scientific Chemicals
J6002603
1 g
Each for $1,493.74
Only null left
 
Description

Description

Peroxidase, horseradish is used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody. Further, it is highly reactive toward human tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lyophilized
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-668-6

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only