Learn More
Brady™ PermaSleeve™ Heat-shrink Labels
Permanently mark wires with the durable PermaSleeve™ PS Heat-shrink polyolefin sleeve label material (B-342). Designed for use with a durable Brady™ handheld label printer.
$75.43 - $167.92
Specifications
| Certifications/Compliance | UL Recognised to UL224;SAE-AMS-DTL-23053/5 C (Class 1);SAE AS-81531 Compliant with Directive 2011/65/EU and 2015/863 |
|---|---|
| Adhesive Type | Non-adhesive |
| Finish | Matte |
| Material | Polyolefin |
| Sterility | Non-sterile |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Format | Color | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Format | Color | Price | Quantity & Availability | |||
|
17012114
|
Brady™
M4C125342 |
5.97 mm (H) x 2.13 m (L) | Continuous | White |
Each for $75.43
|
|
|||
|
17012183
|
Brady™
M4C125342YL |
5.97 mm (H) x 2.13 m (L) | Continuous | Yellow |
Each for $90.03
|
|
|||
|
17012235
|
Brady™
M4125075342 |
19.05 mm (W) x 5.97 mm (H) | Die-cut | White |
Each for $88.82
|
|
|||
|
17012244
|
Brady™
M51251342 |
25.40 mm (W) x 5.97 mm (H) | Die-cut | White |
Each for $88.82
|
|
|||
|
17012327
|
Brady™
M4C187342 |
8.51 mm (H) x 2.13 m (L) | Continuous | White |
Each for $82.74
|
|
|||
|
17012153
|
Brady™
M4C187342YL |
8.51 mm (H) x 2.13 m (L) | Continuous | Yellow |
Each for $111.95
|
|
|||
|
17012329
|
Brady™
M4187075342 |
19.05 mm (W) x 8.51 mm (H) | Die-cut | White |
Each for $97.33
|
|
|||
|
17012324
|
Brady™
M51871342 |
25.40 mm (W) x 8.51 mm (H) | Die-cut | White |
Each for $93.69
|
|
|||
|
17012196
|
Brady™
M4C250342 |
11.15 mm (H) x 2.13 m (L) | Continuous | White |
Each for $93.69
|
|
|||
|
17012283
|
Brady™
M4C250342YL |
11.15 mm (H) x 2.13 m (L) | Continuous | Yellow |
Each for $93.69
|
|
|||
|
17012170
|
Brady™
M4250075342 |
19.05 mm (W) x 11.15 mm (H) | Die-cut | White |
Each for $119.25
|
|
|||
|
17012337
|
Brady™
M52501342 |
25.78 mm (W) x 11.15 mm (H) | Die-cut | White |
Each for $113.17
|
|
|||
|
17012201
|
Brady™
M4C375342 |
16.38 mm (H) x 2.13 m (L) | Continuous | White |
Each for $119.25
|
|
|||
|
17012252
|
Brady™
M4375075342 |
19.05 mm (W) x 16.38 mm (H) | Die-cut | White |
Each for $167.92
|
|
|||
|
17012120
|
Brady™
M53751342 |
25.40 mm (W) x 16.38 mm (H) | Die-cut | White |
Each for $163.05
|
|
|||
Description
- Designed specifically for rigorous cable and wire marking applications
- Self-extinguishing and resistant to extreme temperatures, UV light, chemicals, fuel and oil
- Fast shrinking to save installation time and cost
- Made with a crisp, clear identification surface that is resistant to abrasions and fading
Specifications
| UL Recognised to UL224;SAE-AMS-DTL-23053/5 C (Class 1);SAE AS-81531 Compliant with Directive 2011/65/EU and 2015/863 | |
| Matte | |
| Non-sterile | |
| -55°C to 135°C |
| Non-adhesive | |
| Polyolefin | |
| -67°F to 275°F |