missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periodic Acid, Reagent, 99%, Spectrum™ Chemical
P1035, 10450-60-9, H5IO6
Supplier: Spectrum Chemical Mfg Cor P103525KG
Description
Spectrum™ Chemical Periodic Acid, ReagentSpecifications
| 10450-60-9 | |
| Amber Glass Bottle | |
| H5IO6 | |
| TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
| tetrahydroxyiodous acid | |
| 99% |
| 1 | |
| 2.5 kg | |
| MFCD00011344 | |
| O[I](O)(O)(O)(O)=O | |
| 227.94 | |
| Reagent |
Chemical Identifiers
| 10450-60-9 | |
| 227.94 | |
| TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
| O[I](O)(O)(O)(O)=O |
| H5IO6 | |
| MFCD00011344 | |
| tetrahydroxyiodous acid |