missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, Reagent, 99%, Spectrum™ Chemical

Catalog No. p-7157963
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605034 25 g Amber Glass Bottle
18605036 500 g Amber Glass Bottle
18605037 2.5 kg Amber Glass Bottle
18605038 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605034 Supplier Spectrum Chemical Mfg Cor Supplier No. P103525GM
Only null left

P1035, 10450-60-9, H5IO6

Spectrum™ Chemical Periodic Acid, Reagent

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
IUPAC Name tetrahydroxyiodous acid
SMILES O[I](O)(O)(O)(O)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 99%
Grade Reagent