accessibility menu, dialog, popup

UserName

OHIHOOHOHHOO

CAS RN 10450-60-9

IUPAC Name: tetrahydroxyiodous acid
Synonyms: acid, periodic pentahydroxidooxidoiodine orthoperiodic acid acid, paraperiodic paraperiodic acid periodic acid (HIO4) acids, periodic periodic acid (H5IO6) periodic acid (hio4) hexaoxoiodic(5-) acid
Molecular Weight (g/mol): 227.94
Molecular Formula: H5IO6
InChi Key: TWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILES: O[I](O)(O)(O)(O)=O
Quantity 
  • (6)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (5)
Grade 
  • (1)
  • (3)
  • (2)
  • (3)
  • (7)
  • (3)
Percent Purity 
  • (5)
  • (3)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (8)
  • (2)
  • (2)
  • (8)
  • (11)
  • (6)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
  • (3)
  • (7)
  • (3)
  • (5)
  • (3)
  • (8)