missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Performance Check Solution, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC 5081PC
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Chlorothalonil | 1897-45-6 | 50ng/mL |
| Methyl dacthal | 1861-32-1 | 50ng/mL |
| delta-BHC | 319-86-8 | 40ng/mL |
| Chlorpyrifos | 2921-88-2 | 2ng/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Performance Check Solution | |
| 2mL amber prescored glass ampule | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| C14H9Cl5 | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| YVGGHNCTFXOJCH-UHFFFAOYSA-N | |
| MTBE/Ethyl Acetate | |
| 354.476 | |
| 3036 |
| USEPA Method 508.1 | |
| 50-29-3 | |
| For US EPA Methods 508, 508.1 | |
| US EPA Method 508 | |
| Certified Reference Material | |
| C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)C(Cl)(Cl)Cl)Cl | |
| 1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene | |
| 1 mL | |
| CHEBI:16130 |
Chemical Identifiers
| 50-29-3 | |
| 354.476 | |
| 3036 | |
| 1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene |
| C14H9Cl5 | |
| YVGGHNCTFXOJCH-UHFFFAOYSA-N | |
| CHEBI:16130 | |
| C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)C(Cl)(Cl)Cl)Cl |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only