accessibility menu, dialog, popup

UserName

CAS RN 50-29-3

ClClClClCl

CAS RN 50-29-3

IUPAC Name: 1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene
Synonyms: detox caswell no. 308 chlorophenothanum benzochloryl aavero-extra epa pesticide chemical code 029201 neocidol (solid) neocidol dichlorodiphenyltrichloroethane (ddt) ddt 50 WP
Molecular Weight (g/mol): 354.48
Molecular Formula: C14H9Cl5
InChi Key: YVGGHNCTFXOJCH-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(C1=CC=C(Cl)C=C1)C(Cl)(Cl)Cl
Solvent or Matrix 
  • (1)
Technique 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results