missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peracetic Acid Solution, Approx. 32%, Spectrum™ Chemical
P1023, C2H4O3
$1142.80 - $18498.83
Chemical Identifiers
| CAS | 79-21-0 |
|---|---|
| Molecular Formula | C2H4O2 |
| Molecular Weight (g/mol) | 60.05 |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| IUPAC Name | acetic acid |
| SMILES | CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18605025
|
Spectrum Chemical Mfg Cor
P1023100ML |
100 mL |
Each for $1,142.80
|
|
|||||
|
18605026
|
Spectrum Chemical Mfg Cor
P1023500ML |
500 mL |
Each for $3,190.89
|
|
|||||
|
18605027
|
Spectrum Chemical Mfg Cor
P10231135KGBL |
113.5 kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||