missing translation for 'onlineSavingsMsg'
Learn More

Peracetic Acid Solution, Approx. 32%, Spectrum™ Chemical

Numéro de catalogue. 18605027
Change view
Click to view available options
Quantity:
100 mL
500 mL
113.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
18605027 113.5 kg
18605025 100 mL
18605026 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 18605027 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur P1023113.5KGBL

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

P1023, C2H4O3

Spectrum™ Chemical Peracetic Acid Solution

Identifiants chimiques

CAS 79-21-0
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Spécifications

CAS Max % 0.355
Packaging Drum
Quantity 113.5 kg
Percent Purity ∼32%
Grade Reagent