missing translation for 'onlineSavingsMsg'
Learn More

Peracetic Acid Solution, Approx. 32%, Spectrum™ Chemical

Catalog No. 18605025
Change view
Click to view available options
Quantity:
100 mL
500 mL
113.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18605025 100 mL
18605026 500 mL
18605027 113.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605025 Supplier Spectrum Chemical Mfg Cor Supplier No. P1023100ML
Only null left

P1023, C2H4O3

Spectrum™ Chemical Peracetic Acid Solution

Chemical Identifiers

CAS 79-21-0
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Max % 0.355
Quantity 100 mL
Percent Purity ∼32%
Grade Reagent