missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pepstatin A, 98%

Catalog No. p-7082222
Change view
Click to view available options
Quantity:
5 mg
25 mg
Product Type:
Pepstatin
Protease Inhibitor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
AAJ60237MB 25 mg Protease Inhibitor
AAJ60237LB0 5 mg Pepstatin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60237MB Supplier Thermo Scientific Chemicals Supplier No. J60237MB
Only null left

Protease inhibitor that acts on pepsin, renin and cathepsin D | CAS: 26305-03-3 | C34H63N5O9, C34H63N5O9 | 685.90 g/mol

Pepstatin A, is used as an irreversible inhibitor of HIV-1, aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d. Pepstatin A also efficiently inhibits HIV-1 protease and Fcγ receptor induction by IFN-γ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26305-03-3

Specifications

For Use With (Application) Protease inhibitor that acts on pepsin, renin and cathepsin D
Molecular Weight (g/mol) 685.90g/mol, 685.90
Purity 98%
Molecular Formula C34H63N5O9, C34H63N5O9
Content And Storage 2°C to 8°C
Beilstein 2201362
Form Powder
InChI Key FAXGPCHRFPCXOO-JHTFMCFMSA-N
SMILES CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O
IUPAC Name 3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid
Quantity 25 mg
Product Type Protease Inhibitor
Show More Show Less
EINECSNumber 247-600-0
Recommended Storage Keep cold

RUO – Research Use Only