missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Pepstatin A, 98%
Description
Pepstatin A, is used as an irreversible inhibitor of HIV-1, aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d. Pepstatin A also efficiently inhibits HIV-1 protease and Fcγ receptor induction by IFN-γ.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| For Use With (Application) | Protease inhibitor that acts on pepsin, renin and cathepsin D |
| Molecular Weight (g/mol) | 685.90 |
| Purity | 98% |
| Molecular Formula | C34H63N5O9 |
| Content And Storage | 2°C to 8°C |
| Beilstein | 2201362 |
| Form | Powder |
| InChI Key | FAXGPCHRFPCXOO-JHTFMCFMSA-N |
| SMILES | CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O |
| IUPAC Name | 3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid |
| Show More |
Safety and Handling
| EINECSNumber | 247-600-0 |
| Recommended Storage | Keep cold |
RUO – Research Use Only