missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pepsin

Catalog No. MP210259820
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210259820 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210259820 Supplier MP Biomedicals™ Supplier No. 0210259820

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice

Pepsin is a peptidase and the major proteolytic enzyme produced in the stomach. It is used to digest proteins through the cleavage of interior peptide linkages. It is commonly used in the preparation of Fab fragments from antibodies. Pepsin from porcine gastric mucosa has been used to hydrolyze dry cervical samples in mice.

  • Pepsin hydrolyzes peptide bonds, not amide or ester linkages
  • Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond
  • Sulfur-containing amino acids increase susceptibility to hydrolysis when they are close to the peptide bond
  • Pepsin preferentially cleaves at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid residues
  • Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin

Specifications

Product Type Protease
CAS 9001-75-6
For Use With (Application) Used as a proteolytic enzyme, Used to break down the proteins into their components, i.e. peptides and aminoacids
Quantity 20 g

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.