missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PepMap™ Neo Trap Cartridge

Catalog No. 03255018
Change view
Click to view available options
Length (Metric):
5 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
03255018 5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03255018 Supplier Thermo Scientific™ Supplier No. 174500

Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge with PepMap™ Neo UHPLC Columns to filter impurities from your peptide mapping samples.

Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge with Double nanoViper™ PepMap™ Neo UHPLC Columns or EASY-Spray™ PepMap™ Neo UHPLC Columns to filter impurities from your peptide mapping samples. Connect the trap cartridge to your HPLC system with the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings (Catalog Numbers 174501 and 174502).

Specifications

Column Format Trap Cartridge
Diameter (Metric) 300 μm
For Use With PepMap™ Neo Nano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings (part number 174502)
Length (Metric) 5 mm
Max. Pressure 22,000 psi (1500 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 100 Å
Surface Area 450 m2/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line PepMap Neo
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Description PepMap™ Neo 5 μm C18 300 μm X 5 mm Trap Cartridge
For Use With (Application) Trap and Elute
Show More Show Less