missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PEP 005

Catalog No. 40541
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40541 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40541 Supplier R&D Systems™ Supplier No. 4054/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Protein kinase C activator

Activator of protein kinase C (PKC). Exhibits antileukemic activity; induces nuclear translocation of PKCδ, and induces apoptosis in acute myeloid leukemia (AML) cell lines and primary AML blast cells. Also displays antiproliferative effects and induces apoptosis in Colo205 cells. Reactivates latent HIV in cell culture. Also acts as a substrate for multidrug transporter.

Chemical Identifiers

CAS 75567-37-2
Molecular Formula C25H34O6

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 430.53
Purity >98%
Molecular Weight (g/mol) 430.54
Chemical Name or Material PEP 005
Recommended Storage Store at -20°C