missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™
$192.00 - $480.92
Chemical Identifiers
| CAS | 13642-97-2 |
|---|---|
| Molecular Formula | C10H5F5O2 |
| Molecular Weight (g/mol) | 252.14 |
| MDL Number | MFCD00042332 |
| InChI Key | NIJWSVFNELSKMF-UHFFFAOYSA-N |
| Synonym | Methacrylic Acid Pentafluorophenyl Ester |
| PubChem CID | 193480 |
| IUPAC Name | (2,3,4,5,6-pentafluorophenyl) 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F |
Chemical Identifiers
| 13642-97-2 | |
| 252.14 | |
| NIJWSVFNELSKMF-UHFFFAOYSA-N | |
| 193480 | |
| CC(=C)C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F |
| C10H5F5O2 | |
| MFCD00042332 | |
| Methacrylic Acid Pentafluorophenyl Ester | |
| (2,3,4,5,6-pentafluorophenyl) 2-methylprop-2-enoate |
Specifications
| 13642-97-2 | |
| C10H5F5O2 | |
| 1 g | |
| NIJWSVFNELSKMF-UHFFFAOYSA-N | |
| (2,3,4,5,6-pentafluorophenyl) 2-methylprop-2-enoate | |
| 193480 | |
| ≥97.0% (GC) | |
| Pentafluorophenyl Methacrylate (stabilized with MEHQ) |
| Yellow | |
| MFCD00042332 | |
| Methacrylic Acid Pentafluorophenyl Ester | |
| CC(=C)C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F | |
| 252.14 | |
| 252.14 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator