accessibility menu, dialog, popup

UserName

H3CCH2OOFFFFF

CAS RN 13642-97-2

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-methylprop-2-enoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 252.14
Molecular Formula: C10H5F5O2
InChi Key: NIJWSVFNELSKMF-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results