missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl Chlorothionoformate 95.0+%, TCI America™

Catalog No. P12675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12675G 5 g
P12671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12675G Supplier TCI America Supplier No. P12675G
Only null left

Chemical Identifiers

CAS 135192-53-9
Molecular Formula C7ClF5OS
Molecular Weight (g/mol) 262.578
MDL Number MFCD00075405
InChI Key DKFQHZNKNWNZCO-UHFFFAOYSA-N
Synonym Chlorothioformic Acid O-Pentafluorophenyl Ester, Pentafluorophenyl Thionochloroformate, O-Perfluorophenyl Chlorothioformate
PubChem CID 5214725
IUPAC Name O-(2,3,4,5,6-pentafluorophenyl) chloromethanethioate
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=S)Cl

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 262.58
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluorophenyl Chlorothionoformate