accessibility menu, dialog, popup

UserName

CAS RN 135192-53-9

FFFFFOSCl

CAS RN 135192-53-9

IUPAC Name: 2,3,4,5,6-pentafluorophenyl chloromethanethioate
Synonyms: 2,3,4,5,6-pentafluorophenyl chloromethanethioate
Molecular Weight (g/mol): 262.58
Molecular Formula: C7ClF5OS
InChi Key: DKFQHZNKNWNZCO-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(Cl)=S)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results